Anti-DDIT4L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6376R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DNA-damage-inducible transcript 4-like
- Antigen symbol:DDIT4L
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:115265
- Antigen synonyms:Rtp801L|DDIT4L|REDD-2|HIF-1 responsive protein RTP801-like|REDD2|Protein regulated in development and DNA damage response 2|DNA damage-inducible transcript 4-like protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:150-193/193
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DDIT4L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat