Anti-PPAT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-6359R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Phosphoribosyl Pyrophosphate Amidotransferase
- Antigen symbol:PPAT
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5471
- Antigen synonyms:ATase|EC 2.4.2.14|PPAT|Glutamine phosphoribosylpyrophosphate amidotransferase|GPAT|Amidophosphoribosyltransferase [Precursor]
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:350-400/517
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Phosphoribosyl pyrophosphate amidotransferase (PPAT) is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. The genes for PPAT and PAICS/AIRC, (a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway), are located in close proximity on chromosome 4.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PPAT
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat