Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GABBR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-GABBR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5356R-A488
Supplier:  Bioss
Anti-GABBR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5356R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gamma-Aminobutyric Acid (GABA) B Receptor, 2
  • Antigen symbol:
    GABBR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9568
  • Antigen synonyms:
    GABABR2|HRIHFB2099|GPRC3B|GPR51|HG20
  • Modification:
    phospho-Ser783
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GABBR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat