Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CYP39A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-CYP39A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Katalogové číslo BOSSBS-5050R-A350
Dodavatel:  Bioss
Anti-CYP39A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Katalogové číslo BOSSBS-5050R-A350
Dodavatel:  Bioss

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome P450, Family 39, Subfamily A, Polypeptide 1
  • Antigen symbol:
    CYP39A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51302
  • Antigen synonyms:
    Oxysterol 7 alpha hydroxylase|cytochrome P450 subfamily XXXIX oxysterol 7 alpha hydroxylase polypeptide 1 |cytochrome P450 family 39|oxysterol 7alpha hydroxylase|24 hydroxycholesterol 7 alpha hydroxylase|CP39A_HUM╦?|subfamily A polypeptide 1|hCYP39A1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifikace

O tomto produktu

The CYP39A1 gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of steroids, cholesterol and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP39A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: