Anti-APOC2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-5036R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Apolipoprotein C2
- Antigen symbol:APOC2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:344
- Antigen synonyms:MGC75082|APO-CII|APOC-II
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Apolipoprotein C-II (apoCII) is in found in chylomicrons (large lipoprotein particles absorbed from the gastrointestinal tract) and VLDL (large lipoproteins that are broken down to eventually form LDL). ApoCII activates the enzyme lipoprotein lipase, which hydrolyzes triglycerides and thus provides free fatty acids for cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat