Anti-XPB Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-4260R-CY3
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Xeroderma Pigmentosum, complementation group B
- Antigen symbol:XPB
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2071
- Antigen synonyms:xeroderma pigmentosum|DNA repair protein complementing XP-B cells|basic transcription factor 2 89 kDa subunit|GTF2H|BTF2|xeroderma pigmentosum group B-complementing protein|DNA excision repair protein ERCC-3|TFIIH basal transcription factor complex helicase XPB subunit|complementation group B|XPB|BTF2 p89|TFIIH|TFIIH p89|TFIIH 89 kDa subunit|TFIIH basal transcription factor complex 89 kDa subunit|RAD25
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:625-675/782
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
ATP-dependent 3'-5' DNA helicase, component of the core-TFIIH basal transcription factor, involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. Acts by opening DNA either around the RNA transcription start site or the DNA damage.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: XPB
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat