- Antibody type:Primary
- Antigen name:Oxidative Stress Responsive 1
- Antigen symbol:OXSR1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9943
- Antigen synonyms:OXSR 1|KIAA1101|Oxidative stress responsive 1 protein|OSR1|OSR 1|Serine/threonine protein kinase OSR1.|Oxidative stress responsive 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
OXSR1 is a serine/threonine kinase which regulates downstream kinases in response to environmental stress such as osmotic stresses, notably sorbitol and, to a lesser extent, NaCl. OXSR1 phosphorylated thr84 within the N-terminal regulatory domain of PAK1. Replacement of thr84 with gln reduced activation of PAK1 by an active form of the small G protein CDC42, suggesting that phosphorylation by OXSR1 modulates the G protein sensitivity of PAK. OXSR1 interacts with chloride channel proteins SLC12A6 isoform 2, SLC12A1 and SLC12A2 but not with SLC12A4 and SLC12A7, possibly establishing sensor/signaling modules that initiate the cellular response to environmental stress. Binds to and phosphorylates RELL1, RELL2 AND RELT. OXSR1 may have a role in regulating the actin cytoskeleton.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: OXSR1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat