Anti-GPR15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-3699R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:G Protein-coupled Receptor 15
- Antigen symbol:GPR15
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2838
- Antigen synonyms:GPR15|BOB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:91-120/360
- Purification:Purified by Protein A
- Pk:100 µl
GPR15 is a probable chemokine receptor,its expression has been reported in CD4(+) T lymphocytes, alveolar macrophages, and the basal surface of the small intestinal epithelium. ESTs have been isolated from normal brain and kidney cancer libraries.This gene encodes a G protein-coupled receptor that acts as a chemokine receptor for human immunodeficiency virus type 1 and 2. The encoded protein localizes to the cell membrane. [provided by RefSeq, Nov 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPR15
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat