Order Entry
Czech Republic
ContactUsLinkComponent
Anti-14-3-3 delta+zeta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-14-3-3 delta+zeta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-3000R-FITC
Supplier:  Bioss
Anti-14-3-3 delta+zeta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-3000R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    14-3-3 delta + zeta
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7534, 7529, 10971
  • Antigen synonyms:
    FAS|KCIP-1|Factor activating exoenzyme S|Protein kinase C inhibitor protein-1|14-3-3 protein zeta/delta
  • Modification:
    phospho-Ser58
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    30-70/245
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Members of the 14-3-3 family of proteins are highly conserved proteins, localized in neurons, and are axonally transported to the nerve terminals. They are also present, at lower levels, in various other eukaryotic tissues. 14-3-3 proteins appear to play important roles in a variety of signal transduction pathways, including those involved in cell cycle regulation and cell survival. Because 14-3-3 proteins bind to specific phosphoserine-containing sequences they are likely to have an important role in signaling pathways mediated by serine/threonine protein kinases. Evidence indicates 14-3-3 is required for Raf 1 kinase activity and phosphorylation amoung many other functions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 14-3-3 δ + ζ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat