Anti-PDSS2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-10178R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Prenyl (decaprenyl) Diphosphate Synthase, Subunit 2
- Antigen symbol:PDSS2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Dog
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:DLP1|bA59I9.3|hDLP1|COQ10D3|C6orf210
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
The protein encoded by this gene is an enzyme that synthesizes the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. Defects in this gene are a cause of coenzyme Q10 deficiency.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDSS2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog