- Antibody type:Primary
- Antigen name:Tyrosinase
- Antigen symbol:TYR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7299
- Antigen synonyms:Tumor rejection antigen AB|ATN|CMM8|SHEP3|Monophenol monooxygenase|OCAIA|OCA1A|OCA1|TYR|LB24-AB|SK29-AB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Tyrosinase is the key enzyme for melanin,synthesis in mammalian melanocytes and has been considered to be a unique marker for the study of melanocyte differentiation. A cDNA library was constructed from poly(A)+ mRNA from mouse melanocytes and screened using anti- tyrosinase antiserum and oligonucleotide probes corresponding to amino acid sequence of tyrosinase. sequencing of some cDNA clones positive in these screenings gave a nucleotide sequence of 1838 nucleotides including a open reading frame of 1344 nucleotides that was found to correspond exactly to the amino acid sequence of the cyanogen bromide fragments of tyrosinase.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TYR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat