Anti-SULF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-3894R-A488
: Bioss
- Druh protilátky:Primární
- Název antigenu:Sulfatase 2
- Symbol antigenu:SULF2
- Klonovalita:Polyclonal
- Spojení:Alexa Fluor® 488
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Křížová adsorbce:No
- Formulář:liquid
- Identifikační číslo genu:55959
- Synonyma antigenu:KIAA1247|FLJ44488|MGC126411|HSULF-2|DKFZp313E091|RP5-1049G16.1|FLJ90554
- Skladovací pufr:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Skladovací teplota:Store at 4°C for 12 months
- Koncentrace:1 μg/μl
- Transportní teplota:4°C
- Imunogen:680-730/870
- Čištění:Purified by Protein A
- Bal.:100 µl
Exhibits arylsulfatase activity and highly specific endoglucosamine-6-sulfatase activity. It can remove sulfate from the C-6 position of glucosamine within specific subregions of intact heparin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SULF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat