Anti-RPS6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-3565R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein S6
- Antigen symbol:RPS6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:6194
- Antigen synonyms:Phosphoprotein NP33|RPS6|40S ribosomal protein S6|S6|OK/SW-cl.2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:180-220/249
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May play an important role in controlling cell growth and proliferation through the selective translation of particular classes of mRNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RPS6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat