Anti-RELB Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-3562R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:V-rel Avian Reticuloendotheliosis Viral Oncogene homolog B
- Antigen symbol:RELB
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5971
- Antigen synonyms:RELB|v-rel reticuloendotheliosis viral oncogene homolog B|v-rel reticuloen|v-rel avian reticuloendotheliosis viral oncogene homolog B (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3)|Transcription factor RelB|IREL|I-Rel|REL-B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The NFKB complex consists of NFKB1 or NFKB2 bound to REL, RELA, or RELB. The NFKB complex is inhibited by I kappa B proteins (NFKBIA, or NFKBIB), which inactivate NF kappa B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I kappa B proteins by kinases (IKBKA, or IKBKB,) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF kappa B complex. For some genes, activation requires NFKB interaction with other transcription factors, such as STAT, AP1 (JUN), and NFAT.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RELB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat