Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RCC1 Rabbit Polyclonal Antibody (Cy5®)
Anti-RCC1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3545R-CY5
Supplier:  Bioss
Anti-RCC1 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-3545R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Regulator of Chromosome Condensation 1
  • Antigen symbol:
    RCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1104
  • Antigen synonyms:
    CHC1|SNHG3-RCC1|RCC1-I
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    2-50/421
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Ran GTPase plays important roles in nucleocytoplasmic transport in interphase and in both spindle formation and nuclear envelope (NE) assembly during mitosis. The latter functions rely on the presence of high local concentrations of GTP bound Ran near mitotic chromatin. RanGTP localization has been proposed to result from the association of Ran's GDP/GTP exchange factor, RCC1, with chromatin , but Ran is shown here to bind directly to chromatin in two modes, either dependent or independent of RCC1, and, where bound, to increase the affinity of chromatin for NE membranes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RCC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat