Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CRX Rabbit Polyclonal Antibody (Cy5.5®)
Anti-CRX Rabbit Polyclonal Antibody (Cy5.5®)
  BOSSBS-3798R-CY5.5
 :  Bioss
Anti-CRX Rabbit Polyclonal Antibody (Cy5.5®)
  BOSSBS-3798R-CY5.5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cone-rod Homeobox
  • Antigen symbol:
    CRX
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    1406
  • Antigen synonyms:
    LCA7|OTX3|CRD|CORD2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    60-110/299
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Transcription factor that binds and transactivates the sequence 5'-TAATC[CA]-3' which is found upstream of several photoreceptor-specific genes, including the opsin genes. Acts synergistically with other transcription factors, such as NRL, RORB and RAX, to regulate photoreceptor cell-specific gene transcription. Essential for the maintenance of mammalian photoreceptors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: