Anti-MPG Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-2412R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:N-Methylpurine DNA Glycosylase
- Antigen symbol:MPG
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4350
- Antigen synonyms:MPG|3' end of the Mid1 gene|CRA36.1|localized 68 kb upstream the humanzeta globin gene on 16p|AAG|proliferation-inducing protein 16|Mid1|alkyladenine DNA glycosylase|MDG|PIG11|N-methylpurine-DNA glycosylase|anpg|proliferation-inducing protein 11|APNG|CRA36.1 (3-methyl-adenine DNA glycosylase)|PIG16
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
MPG (N-methylpurine DNA glycosylase, or 3-methyladenine (3MeA) DNA glycosylase) repairs the 3MeA lethal lesion blocking DNA replication in Escherichia coli. Analysis of human cDNA libraries found a cDNA that would hybridize to human genomic DNA but not to E. coli or yeast DNA. This MPG gene has been mapped to human chromosome 16 by analysis of a panel of DNAs from mouse/human and hamster/human hybrid cell lines.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MPG
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat