Anti-GPAT4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1924R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Glycerol-3-phosphate Acyltransferase 4
- Antigen symbol:GPAT4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:137964
- Antigen synonyms:LPAATZ|1-AGPAT 6|LPAAT-zeta|AGPAT6|TSARG7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
AGPAT6 is a novel glycerolipid acyltransferase of the ER, which is crucial for the production of milk fat by the mammary gland. It converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSARG7
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat