Anti-GPR78 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1807R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:G Protein-coupled Receptor 78
- Antigen symbol:GPR78
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:27201
- Antigen synonyms:G-protein coupled receptor 78|UNQ5925/PRO19818|GPR78
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:240-290/363
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Orphan receptor. Displays a significant level of constitutive activity. Its effect is mediated by G(s)-alpha protein that stimulate adenylate cyclase, resulting in an elevation of intracellular cAMP.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPR78
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human