Specifications
- Antibody type:Primary
- Antigen name:muskelin 1, intracellular mediator containing kelch motifs
- Antigen symbol:MKLN1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:TWA2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Muskelin is a 735 amino acid nuclear and cytoplasmic protein that mediates cytoskeletal responses and cell spreading to Thrombospondin 1, an extracellular matrix glycoprotein. Forming a CTLH complex with RMND5A, Ran BP-M and TWA1, Muskelin exists as two alternatively spliced isoforms. Muskelin is implicated as an isoform-specific anchoring protein for the prostaglandin EP3 receptor, and contains one LisH domain, a CTLH domain, six Kelch repeats and an N-terminal discoidin-like domain through which it self-associates via a head-to-tail mechanism. Human and mouse Muskelin share 98% amino acid seqeuence homology and are encoded by genes located on chromosomes 7q32.3 and 6 A3.3, respectively.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MKLN1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat