Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-ADGRG7 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-13517R-CY7
 :  Bioss
Anti-ADGRG7 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-13517R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Adhesion G Protein-Coupled Receptor G7
  • Antigen symbol:
    ADGRG7
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    84873
  • Antigen synonyms:
    GPR128
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

G protein-coupled receptors (GPCRs), also designated seven transmembrane (7TM) receptors and heptahelical receptors, are a protein family which interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers such as diacylglycerol, cyclic AMP, inositol phosphates, and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. G protein receptor 128 (GPR128), a member of the secretin family of GCPRs with a GPS domain in its N-terminal domain, may mediate signaling processes to the interior of the cell via activation of G proteins. GPR128 represents an allopeptide which may be involved in T cell mediated transplant rejection as it is able to stimulate 2.102 T cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRG7
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human