Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-TRP73 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-1346R-CY5
 :  Bioss
Anti-TRP73 Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-1346R-CY5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    transformation related protein 73
  • Antigen symbol:
    TRP73
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    22062
  • Antigen synonyms:
    p73|Tp73|TAp73|deltaNp73
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    590-625/631
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRP73
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat