Anti-CFD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Katalogové číslo BOSSBS-13130R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Complement Factor D
- Antigen symbol:CFD
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1675
- Antigen synonyms:ADIPSIN|ADN|PFD|DF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:20-70/253
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Factor D cleaves factor B when the latter is complexed with factor C3b, activating the C3bbb complex, which then becomes the C3 convertase of the alternate pathway. Its function is homologous to that of C1s in the classical pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CFD
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: