Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box I1
- Antigen symbol:FOXI1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:2299
- Antigen synonyms:FKHL10|HFH-3|FREAC6|HFH3|FKH10|FREAC-6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FOXI1 is a member of the FOX family of transcription factors. The FOX family is a large group of proteins (consisting of at least 43 members) that share a common DNA binding domain termed winged-helix or forkhead domain. FOX transcription factors play important roles in development, differentiation, aging and hormone responsiveness. Localizing to the nucleus, FOXI1 functions as a transcription factor. Mice with mutated forms of FOXI1 show defects in ear development, implying that FOXI1 plays a significant role in the developmental pathway of ears and, in particular, the cochlea and vestibulum. FOXI1 is an upstream transcription regulator of Pendrin (a protein associated with deafness), suggesting a role for FOXI1 in the pathogenesis of Pendred syndrome (PS), a condition of nonsyndromic hearing loss and enlarged vestibular aqueduct (EVA).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: FOXI1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat