Order Entry
Czech Republic
ContactUsLinkComponent
Anti-FOXI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FOXI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13203R-FITC
Supplier:  Bioss
Anti-FOXI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13203R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Forkhead Box I1
  • Antigen symbol:
    FOXI1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2299
  • Antigen synonyms:
    FKHL10|HFH-3|FREAC6|HFH3|FKH10|FREAC-6
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

FOXI1 is a member of the FOX family of transcription factors. The FOX family is a large group of proteins (consisting of at least 43 members) that share a common DNA binding domain termed winged-helix or forkhead domain. FOX transcription factors play important roles in development, differentiation, aging and hormone responsiveness. Localizing to the nucleus, FOXI1 functions as a transcription factor. Mice with mutated forms of FOXI1 show defects in ear development, implying that FOXI1 plays a significant role in the developmental pathway of ears and, in particular, the cochlea and vestibulum. FOXI1 is an upstream transcription regulator of Pendrin (a protein associated with deafness), suggesting a role for FOXI1 in the pathogenesis of Pendred syndrome (PS), a condition of nonsyndromic hearing loss and enlarged vestibular aqueduct (EVA).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FOXI1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat