Specifications
- Antibody type:Primary
- Antigen name:Pre-mRNA Processing Factor 40A
- Antigen symbol:PRPF40A
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:55660
- Antigen synonyms:FNBP3|HYPA|NY-REN-6|FLAF1|HIP-10|Prp40|FBP11|HIP10|FBP-11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Fas ligand associated factor 1; Fas ligand-associated factor 1; FBP 11; FBP11; FLAF 1; FLAF1; FNBP 3; Formin binding protein 11; Formin binding protein 3; Formin-binding protein 11; Formin-binding protein 3; HIP-10; HIP10; Huntingtin interacting protein A; Huntingtin interacting protein HYPA; Huntingtin yeast partner A; Huntingtin-interacting protein 10; Huntingtin-interacting protein A; HYPA; NY REN 6; NY REN 6 antigen; PR40A_HUMAN; Pre mRNA processing factor 40 homolog A; Pre-mRNA-processing factor 40 homolog A; PRP40 pre mRNA processing factor 40 homolog A (S cerevisiae); PRP40 pre mRNA processing factor 40 homolog A (yeast); PRP40 pre mRNA processing factor 40 homolog A; PRPF 40A; PRPF40A; Renal carcinoma antigen NY-REN-6.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRPF40A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat