Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PRPF40A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-13190R-A350
 :  Bioss
Anti-PRPF40A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-13190R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Pre-mRNA Processing Factor 40A
  • Antigen symbol:
    PRPF40A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    55660
  • Antigen synonyms:
    FNBP3|HYPA|NY-REN-6|FLAF1|HIP-10|Prp40|FBP11|HIP10|FBP-11
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Fas ligand associated factor 1; Fas ligand-associated factor 1; FBP 11; FBP11; FLAF 1; FLAF1; FNBP 3; Formin binding protein 11; Formin binding protein 3; Formin-binding protein 11; Formin-binding protein 3; HIP-10; HIP10; Huntingtin interacting protein A; Huntingtin interacting protein HYPA; Huntingtin yeast partner A; Huntingtin-interacting protein 10; Huntingtin-interacting protein A; HYPA; NY REN 6; NY REN 6 antigen; PR40A_HUMAN; Pre mRNA processing factor 40 homolog A; Pre-mRNA-processing factor 40 homolog A; PRP40 pre mRNA processing factor 40 homolog A (S cerevisiae); PRP40 pre mRNA processing factor 40 homolog A (yeast); PRP40 pre mRNA processing factor 40 homolog A; PRPF 40A; PRPF40A; Renal carcinoma antigen NY-REN-6.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRPF40A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat