- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 7
- Antigen symbol:CCR7
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:12775
- Antigen synonyms:Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1|MIP-3 Beta Receptor |EBI1|CCR-7|Chemokine (C-C Motif) Receptor 7|CD197|CMKBR7
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:25-59/379
- Purification:Purified by Protein A
- Pk:100 µl
CCR7 is a member of the G protein coupled receptor family (subfamily : chemokine). This receptor was identified as a gene induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. It is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCR7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat