Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CCR7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1305R-A350
 :  Bioss
Anti-CCR7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1305R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Chemokine (C-C motif) receptor 7
  • Antigen symbol:
    CCR7
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    12775
  • Antigen synonyms:
    Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1|MIP-3 Beta Receptor |EBI1|CCR-7|Chemokine (C-C Motif) Receptor 7|CD197|CMKBR7
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    25-59/379
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

CCR7 is a member of the G protein coupled receptor family (subfamily : chemokine). This receptor was identified as a gene induced by the Epstein Barr virus (EBV), and is thought to be a mediator of EBV effects on B lymphocytes. CCR7 has been reported to be expressed in blood, bone marrow, lymph node, and intestine. It is particularly expressed in lymphoid tissues and in activated B and T lymphocytes and has been shown to control the migration of memory T cells to inflamed tissues, as well as stimulate dendritic cell maturation. The chemokine (C-C motif) ligand 19 (CCL19/ECL) has been reported to be a specific ligand of this receptor. ESTs have been isolated from blood, embryo, lymph node, and thymus libraries.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCR7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat