Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-POLK Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-13020R-CY5
 :  Bioss
Anti-POLK Rabbit Polyclonal Antibody (Cy5®)
  BOSSBS-13020R-CY5
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Polymerase (DNA Directed) Kappa
  • Antigen symbol:
    POLK
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    27343
  • Antigen synonyms:
    OTTHUMP00000020323|BETAN|BETA-N|POLL|DNA polymerase beta-2|pol lambda|polymerase (DNA-directed)|pol beta2|DNA-directed DNA polymerase lambda|POLKAPPA|polymerase (DNA directed)|DNA polymerase kappa (DNA polymerase beta-N)|POL-KAPPA|DNA polymerase lamda2|lambda|FLJ46002|RP11-529I10.1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

DNA polymerase lambda (pol lambda), also designated DNA polymerase lambda or Pol Beta2, is a low-fidelity polymerase which plays a role in both spontaneous and DNA damage-induced mutagenesis. Encoded by the POLL gene, pol lambda is a member of the DNA polymerase type-X family. Pol lambda extends primer-terminal mispairs opposite nondamaged DNA templates, suggesting that it may assist in extending mismatched base pairs during normal DNA replication. In addition, pol ?may play a role in the mutagenic bypass of T-T dimers. Proliferating cell nuclear antigen (PCNA), a protein essential to DNA replication, interacts with pol lambda and thus influences the ability of pol ?to synthesize DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POLK
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat