Anti-RBFOX2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-12299R-A647
: Bioss
- Antibody type:Primary
- Antigen name:RNA Binding Motif Protein 9
- Antigen symbol:RBFOX2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23543
- Antigen synonyms:fxh|RTA|HNRBP2|Fox-2|FOX2|RBM9|HRNBP2|dJ106I20.3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-150/390
- Purification:Purified by Protein A
- Pk:100 µl
RNA-binding protein that regulates alternative splicing events by binding to 5'-UGCAUGU-3' elements. Prevents binding of U2AF2 to the 3'-splice site. Regulates alternative splicing of tissue-specific exons and of differentially spliced exons during erythropoiesis (By similarity). RNA-binding protein that seems to act as a coregulatory factor of ER-alpha.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RBFOX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat