Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
Anti-CK1g2/Casein Kinase 1 gamma 2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-12425R-CY7
Supplier:  Bioss
Anti-CK1g2/Casein Kinase 1 gamma 2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-12425R-CY7
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    CK1g2/Casein Kinase 1 gamma 2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Cy7®
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at 4°C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. They can phosphorylate a large number of proteins. Participates in Wnt signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CK1g2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity: