Specifications
- Antibody type:Primary
- Antigen name:Tetraspanin 32
- Antigen symbol:TSPAN32
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:10077
- Antigen synonyms:PHMX|tumor-suppressing subchromosomal transferable fragment cDNA 6|pan-hematopoietic expression|MGC22455|FLJ17158|TSSC6|PHEMX|tetraspanin|tetraspanin 32|tumor-suppressing subtransferable candidate 6|ART1|tumor-suppressing STF cDNA 6|FLJ97586|pan-hematopoietic expression protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Phemx is a member of the tetraspanin (TM4SF) family of proteins that may be involved in transmembrane signal transduction, regulation of cell proliferation, differentiation and motility. Phemx is a multi-pass membrane protein containing intracellular N- and C-terminal domains, four transmembrane domains and two extracellular loops. It is ubiquitously expressed from early embryogenesis through adulthood. Phemx exhibits predominant expression in hematopoietic tissues suggesting a role in hematopoietic-cell function. In association with the Integrin αIIb/Integrin β3 complex, Phemx functions to stabilize arterial thrombi in platelets and regulate “outside-in†signaling. This interaction may be important in the process of wound healing. The gene encoding Phemx is located in an important tumor-suppressor gene region that has been associated with Beckwith-Wiedemann syndrome as well as a variety of cancers.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSPAN32
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human