- Antibody type:Primary
- Antigen name:hemogen
- Antigen symbol:HEMGN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:55363
- Antigen synonyms:NDR|EDAG-1|CT155|EDAG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Hemogen is a 484 amino acid protein encoded by the human gene HEMGN. Hemogen is a nuclear protein that is expressed in hematopoietic precursor cells and can be detected in CD34+ and K-562 leukemia cell line. It is also expressed in bone marrow, testis, thymus and thyroid tumors, non-Hodgkin lymphoma, various leukemia cell lines, peripheral blood mononuclear cells (PBMCs) and bone marrow mononuclear cells (BMMCs) of patients with leukemia. Hemogen is down-regulated during megakaryocytic differentiation of K-562 cells by 12-O-tetradecanoylphorbol-13-acetate (TPA) (at protein level). It can be up-regulated in normal PBMCs by mitogens.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HEMGN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human