Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CSN7B Rabbit Polyclonal Antibody (Cy5®)
Anti-CSN7B Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-11540R-CY5
Supplier:  Bioss
Anti-CSN7B Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-11540R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    COP9 Complex Homolog Subunit 7b
  • Antigen symbol:
    CSN7B
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Signalosome subunit 7b.|SGN7b|COP9 Constitutive Photomorphogenic Homolog Subunit 7b|JAB1 Containing Signalosome Subunit 7b|COP9 signalosome complex subunit 7b|COP9 Signalasome Subunit 7b|CSN7B_HUMAN|COPS7B|JAB1-containing signalosome subunit 7b|COP9 Complex Homolog Subunit 7b
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The COP9 signalosome (CSN) complex is involved in several different developmental and cellular processes. The complex is made up of several widely expressed proteins: CSN1 (COPS1), CSN2 (COPS2), CSN3 (COPS3), CSN4 (COPS4), CSN5 (COPS5), CSN6 (COP6), CSN7a (COPS7, COPS7a) or CSN7b (COP7b) and CSN8 (COP8). The CSN complex acts as a regulator for the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes. It is also involved in the phosphorylation of p53, c-Jun, I˚Bå and IRF-8, as well as CSN-dependent phosphorylation of p53, and c-Jun protects and promotes degradation by the Ubl system. CSN7 is phosphorylated by CK2 and is composed of two subunits; a and b. CSN7a contains a PCI (Proteasome CSN9 initiation factor 3) region, as well as a coiled-coil region and is predicted to interact with CSN2, CSN3, CSN4, CSN5, CSN6, CSN8, and GPS1. CSN7b contains only a PCI region and is predicted to interact with INT6.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSN7b
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: