Specifications
- Antibody type:Primary
- Antigen name:Synaptogyrin 4
- Antigen symbol:SYNGR4
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23546
- Antigen synonyms:Synaptogyrin4|Synaptogyrin 4|Synaptogyrin-4|SNG4_HUMAN|MGC125805.|SYNGR4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The synaptogyrin family of proteins are integral membrane proteins containing four transmembrane regions. Synaptogyrins are tyrosine-phosphorylated proteins with two neuronal (Synaptogyrins 1 and 3) and one ubiquitous (Synaptogyrin-2) isoform. Synaptophysin and synaptogyrin represent the major constituents of synaptic vesicles. The 26kDa protein Synaptogyrin-1 is associated with presynaptic vesicles in neuronal cells. Synaptogyrin-2, also known as Cellugyrin has a tyrosine phosphorylated C-terminal cytoplasmic tail, and is involved in the regulation of membrane traffic in non-neuronal cells. Synaptogyrin-3 is expressed mainly in brain and placenta. The SYNGR4 gene encodes for the 234 amino acid protein Synaptogyrin-4.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SYNGR4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat