Order Entry
Czech Republic
ContactUsLinkComponent
Cation exchange chromatography columns, TSK-GEL® SP-NPR
Cation exchange chromatography columns, TSK-GEL® SP-NPR
Catalog # TOYO0013076
Supplier:  TOSOH Bioscience
undefined
Cation exchange chromatography columns, TSK-GEL® SP-NPR
Catalog # TOYO0013076
Supplier:  TOSOH Bioscience

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    4,60 mm
  • Particle Size:
    2,5 µm
  • Pore size:
    -
  • Description:
    Cation exchange chromatography column, SP-NPR, nonporous
  • Length:
    35 mm
  • Column hardware:
    Stainless steel
  • Phase:
    SP-NPR
  • Packing material:
    Polymer
  • Pk:
    1

Specifications

About this item

TSK-Gel® SP-NPR are polymer based stainless steel columns. These columns provide fast separations due to their small spherical particles.

  • Nonporous columns
  • Used in gene therapy research