ContactUsLinkComponent
Size exclusion and ligand exchange chromatography columns, SUGAR KS-802 and KS-801
Size exclusion and ligand exchange chromatography columns, SUGAR KS-802 and KS-801
Supplier:  Resonac
Certificates
undefined
Size exclusion and ligand exchange chromatography columns, SUGAR KS-802 and KS-801
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-2215
  • 554-2214
  • Max. pressure:
  • 5,0 MPa (50 bar)
  • 5,0 MPa (50 bar)
  • pH range:
  • 3 - 7
  • 3 - 7
  • Temperature limit:
  • 20 - 85 °C
  • 20 - 85 °C
  • USP code:
  • L22, L58
  • L22, L58