J.T.Baker® BAKERBOND spe™ C18 PolarPlus®-SDB-1, SPE Columns for Special Applications
7704-06
: Avantor
- Capacity:6 ml
- Description:Column with 250 mg C18 PolarPlus® (top) and 100 mg SDB-1, separated by a PE frit
- Bed weight:350 mg
- Pk:30
These SPE columns are designed for the extraction of polar pesticides from water.