ContactUsLinkComponent
Sporicide, Actril® Cold Sterilant, cleanroom packaging
Sporicide, Actril® Cold Sterilant, cleanroom packaging
Catalog # 148-0115
undefined
Sporicide, Actril® Cold Sterilant, cleanroom packaging
Catalog # 148-0115
Specifications
  • Description:
    Actril® CR, Cold Sterilant
  • Packed:
    1 l
  • Pk:
    6
Specifications
About this item
Actril® Cold Sterilant (Clean Room packaging) is a ready to use, effective, viricide, bactericide, fungicide, and sporicide It is is available in cleanroom packaging to make it easier to use in cleanroom setting. Actril® CR is completely and does not have to be part of a rotation of disinfectants program.
  • No dilution required
  • Faster, safer, and less corrosive than aldehyde-based liquids
  • Filtered through 0,2 micron filters
  • Double bagged and EtO Processed (6 log reduction of B. atrophaeus spores)
  • Complete lot traceability and documentation conforms to the pharma standards
Actril® Cold Sterilant is a leading surface disinfectant in medical and cleanroom applications.
Certifications: Actril® Cold Sterilant complies with the USP guidelines for choosing disinfectants for aseptic areas.
Packaging: 6×1 kg bottles with sprayers per case.