ContactUsLinkComponent
Dehydrated culture media, potato dextrose agar, MP Biomedicals
Dehydrated culture media, potato dextrose agar, MP Biomedicals
Catalog # ICNA091008517
Supplier:  MP Biomedicals
CAS Number:  
Dehydrated culture media, potato dextrose agar, MP Biomedicals
Catalog # ICNA091008517
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    500 g
  • Standards:
    APHA
  • Description:
    Potato dextrose agar, powder
Specifications
About this item
Potato dextrose agar (PDA) is suited for the cultivation of yeasts and moulds from dairy products and other foods. It may also be used to induce sporulation in many fungi.
Certifications: This medium is recommended by the Association of Public Health (APHA).