Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ICNA0855499
Supplier:  MP Biomedicals
Anti-Ig Goat Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ICNA0855499
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulins
  • Antigen symbol:
    Ig
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Concentration:
    Total Protein: 15.9 mg/mL
  • Shipping temperature:
    RT
  • Purification:
    Antibody and FITC are conjugated under defined conditions to obtain the specified molar ratio (3-6 mol/mol). Conjugated protein is purified by gel filtration. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed, and lyophilized.
  • Size:
    2 ml
  • Pk:
    2 mL

Specifications

About this item

The total protein is measured using the Biuret procedure with bovine albumin as the standard; F/P ratio is calculated using absorbance at 491.5nm. Antibody titer of the IgG fraction is standardized with an in-house control by immunoelectrophoresis; No trace of albumin is detected. Antibody activity to non-immunoglobulin serum proteins is not present.

Type: Secondary
Antigen: Ig
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse