Anti-Myc Rabbit Polyclonal Antibody [clone: L56]
BWRLBS1245
- Antibody type:Primary
- Antigen symbol:Myc
- Clonality:Polyclonal
- Clone:L56
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 31-80 of Human Myc.
Myc (L56) pAb detects endogenous levels of proto-oncogene c-Myc protein.
Type: Primary
Antigen: Myc
Clonality: Polyclonal
Clone: L56
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat