Anti-MMP-19 Rabbit Polyclonal Antibody [clone: G44]
Catalog # BWRLBS1235
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-19
- Clonality:Polyclonal
- Clone:G44
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 12-60 of Human MMP-19.
MMP-19 (G44) pAb detects endogenous levels of MMP-19 protein.
Type: Primary
Antigen: MMP-19
Clonality: Polyclonal
Clone: G44
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse