Anti-MMP-13 Rabbit Polyclonal Antibody [clone: L22]
Catalog # BWRLBS1231
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-13
- Clonality:Polyclonal
- Clone:L22
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids N-terminus of Human MMP-13.
MMP-13 (L22) pAb detects endogenous levels of ~60 kDa latent or the ~48 kDa active forms of MMP-13 protein.
Type: Primary
Antigen: MMP-13
Clonality: Polyclonal
Clone: L22
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat