Anti-CK14/16 Rabbit Polyclonal Antibody [clone: S5]
Catalog # BWRLBS1200
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CK14/16
- Clonality:Polyclonal
- Clone:S5
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human Cytokeratin 14
Cytokeratin 14/16 (S5) pAb detects endogenous levels of Cytokeratin 14 and Cytokeratin 16 protein.
Type: Primary
Antigen: CK14/16
Clonality: Polyclonal
Clone: S5
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat