Anti-Cdc6 Rabbit Polyclonal Antibody [clone: V48]
Catalog # BWRLBS1048
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cdc6
- Clonality:Polyclonal
- Clone:V48
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 12-63 of Human Cdc6.
CDC6 (V48) pAb detects endogenous levels of CDC6 protein.
Type: Primary
Antigen: Cdc6
Clonality: Polyclonal
Clone: V48
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat