Anti-SLP-76 Rabbit Polyclonal Antibody
Catalog # BWRLBS7224
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SH2 Domain Containing leukocyte Protein of 76kDa
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human SLP-76.
SLP-76 pAb detects endogenous levels of SLP-76 protein.
Type: Primary
Antigen: SLP-76
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat