Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-p-Connexin 43 Rabbit Polyclonal Antibody [clone: S367]
Catalog # BWRLBS4777
Anti-p-Connexin 43 Rabbit Polyclonal Antibody [clone: S367]
Catalog # BWRLBS4777

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Connexin 43
  • Clonality:
    Polyclonal
  • Clone:
    S367
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human Connexin 43 around the phosphorylation site of Serine 367.

p-Connexin 43 (S367) pAb detects endogenous levels of Connexin 43 protein when phosphorylated at Ser367.

Type: Primary
Antigen: p-Connexin 43
Clonality: Polyclonal
Clone: S367
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat