Anti-AKAP 10 Rabbit Polyclonal Antibody [clone: R27]
Catalog # BWRLBS1877
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AKAP 10
- Clonality:Polyclonal
- Clone:R27
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 1-50 of Human AKAP 10.
AKAP 10 (R27) pAb detects endogenous levels of AKAP 10 protein.
Type: Primary
Antigen: AKAP 10
Clonality: Polyclonal
Clone: R27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat